To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 Kit
- Species reactivity:Rabbit
- Shipping conditions:Wet Ice
- Cat. no.:CAKCA003
- Supplier No.:KCA003
Specifications
About this item
This Chemiluminescent Western Blot Kit allows for the detection of primary Rabbit polyclonal or monoclonal antibodies provided by the user
Recommended Dilutions: Western Blot: YES; Wash buffers MUST NOT contain SODIUM AZIDE or other inhibitors of peroxidase activity!
After protein separation and transfer, the membrane is probed with primary antibody. Detection of the membrane bound primary antibody-antigen complex is achieved by the addition of a secondary antibody conjugated to the enzyme horseradish peroxidase. The enzyme reacts with a specialized formulation of luminol, an extremely sensitive, non-radioactive substrate that emits light and allows visualization using X-ray film or other imaging methods, including highly sensitive CCD cameras and imaging systems. Because of the extremely high sensitivity of our FemtoMax Chemiluminescent Substrate Kit for Western Blotting, primary and secondary antibodies can be used at greater dilutions. If you currently dilute your primary antibody 1:1,000 using an ECL substrate equivalent, then dilute your antibody 1:5,000 using the FemtoMax substrate.