Order Entry
Canada
ContactUsLinkComponent
Chemiluminescent Western Blot Kit for use with Mouse Primary Antibody, FemtoMax™, Rockland Immunochemicals
Chemiluminescent Western Blot Kit for use with Mouse Primary Antibody, FemtoMax™, Rockland Immunochemicals
Catalog # CAKCA002
CAS Number:  
Chemiluminescent Western Blot Kit for use with Mouse Primary Antibody, FemtoMax™, Rockland Immunochemicals
Catalog # CAKCA002
Supplier Number:  KCA002
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 Kit
  • Species reactivity:
    Mouse
  • Shipping conditions:
    Wet Ice
  • Concentration:
    Sufficient to run approximately 30 mini blots (7.5 cm x 8 cm).
  • Cat. no.:
    CAKCA002
  • No. of Assays/Kit:
    Sufficient to run approximately 30 mini blots (7.5×8.0 cm)
  • Supplier No.:
    KCA002

Specifications

About this item

This Chemiluminescent Western Blot Kit allows for the detection of primary mouse polyclonal or monoclonal antibodies provided by the user

Recommended Dilutions: Western Blot: 1:20,000; Wash buffers MUST NOT contain SODIUM AZIDE or other inhibitors of peroxidase activity!

After protein separation and transfer, the membrane is probed with primary antibody. Detection of the membrane bound primary antibody-antigen complex is achieved by the addition of a secondary antibody conjugated to the enzyme horseradish peroxidase. The enzyme reacts with a specialized formulation of luminol, an extremely sensitive, non-radioactive substrate that emits light and allows visualization using X-ray film or other imaging methods, including highly sensitive CCD cameras and imaging systems. Because of the extremely high sensitivity of our FemtoMax Chemiluminescent Substrate Kit for Western Blotting, primary and secondary antibodies can be used at greater dilutions. If you currently dilute your primary antibody 1:1,000 using an ECL substrate equivalent, then dilute your antibody 1:5,000 using the FemtoMax substrate.