Anti-IgG Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
10801-350
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Flow cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Reactivity:Ferret
- Size:1 mg
- Format:IgG
- Form:Lyophilized
- Concentration:1.0 mg/mL by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Ferret IgG gamma heavy chain
- Tested applications:Fluoresence ELISA, Fluorochrome Protein
- Purification:This product was prepared from monospecific antiserum by immunoaffinity chromatography using Ferret IgA coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum, Ferret IgG and Ferret Serum. No reaction was observed against Ferret IgA or Ferret IgM. Specificity was confirmed by ELISA at less than 1% cross reactivity against other Ferret heavy or light chain isotypes.
- Cat. no.:10801-350
- Supplier No.:618-100-012
This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Recommended Dilutions: FLISA: 1:10,000 - 1:50,000; Flow Cytometry: 1:500 - 1:2,500; Fluorochrome Protein Value: 2.0; Immunofluorescence Microscopy: 1:1,000 - 1:5,000 ; contains 0.01% (w/v) Sodium Azide
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Ferret