Specifications
- Description:HBc/HIV/HCV Antibody Test Controls
- Supplier No.:815311006104
Specifications
About this item
HBc/HIV/HCV Antibody Test Controls (Test Controls) are external quality control agents, intended for use with the following Miriad rapid antibody tests:
Model 815311006012 Miriad Rapid HCV/HIV Antibody Test POU+
Model 815311005954 Miriad Rapid HCV/HIV Antibody Test LAB+
Model 815311006005 Miriad Rapid HBc/HIV/HCV Antibody Test POU+
Model 815311005978 Miriad Rapid HBc/HIV/HCV Antibody Test LAB+
Performance may vary if used with other rapid tests.