Order Entry
Canada
ContactUsLinkComponent
Petri Dish, No Stacking Ring, 'Slipable', Mono Plate, Sterile
Petri Dish, No Stacking Ring, 'Slipable', Mono Plate, Sterile
Catalog # CA10753-948
undefined
Petri Dish, No Stacking Ring, 'Slipable', Mono Plate, Sterile
Catalog # CA10753-948
Supplier Number:  3301

Specifications

  • Ø:
    100 mm
  • Sterility:
    Sterile
  • Description:
    Petri dish, no stacking ring, 'slipable', mono plate
  • Height:
    15 mm
  • Cat. no.:
    CA10753-948
  • Packaging:
    500/case
  • Supplier No.:
    3301

Specifications

About this item

Petri dish, with smooth bottom, without stacking ring.

  • Manufactured from pure virgin polystyrene
  • Unsurpassed in optical clarity, permitting unimpaired observation of dish content - ideal for micro photography
  • Engineered for optimum flatness to provide uniform agar thickness - reducing media costs
  • Designed with nesting lugs for optimum culture growth and media shelf life
  • Produced in a wide variety of shapes and sizes

Applications: Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, pharmaceutical manufacturing

Packaging: Petri dishes are packaged 500 per case.