Order Entry
Canada
ContactUsLinkComponent
 
Mouse IgG1 Monoclonal Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # 10753-832
Supplier:  Tonbo Biosciences
undefined
Mouse IgG1 Monoclonal Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # 10753-832
Supplier:  Tonbo Biosciences
Supplier Number:  67-4714-U100

Specifications

  • Antibody type:
    Control
  • Antigen name:
    Immunoglobulin G1
  • Antigen symbol:
    IgG1
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-21
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Size:
    100 µg
  • Cross adsorption:
    No
  • Storage temperature:
    Store undiluted at 2-8°C, protect from light, do not freeze.
  • Cat. no.:
    10753-832
  • Supplier No.:
    67-4714-U100

Specifications

About this item

The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: