Anti-MS4A1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2H7]
Supplier: Tonbo Biosciences
Certificates
About this item
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation.
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 2H7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Specifications
- Catalog No:
- 10753-722
- 10753-720
- 10753-724
- Antigen symbol:
- MS4A1
- MS4A1
- MS4A1
- Antigen name:
- Membrane-Spanning 4-Domains, Subfamily A, Member 1
- Membrane-Spanning 4-Domains, Subfamily A, Member 1
- Membrane-Spanning 4-Domains, Subfamily A, Member 1
- Antigen synonyms:
- CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
- CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
- CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 2H7
- 2H7
- 2H7
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 931
- 931
- 931
- Isotype:
- IgG2b kappa
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Yes
- Cross adsorption:
- No
- No
- No
- Storage temperature:
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Store undiluted at 2-8°C, protect from light, do not freeze.
Specifications
Frequently Bought Together