You searched for: Proteins and Peptides
Proteins are used in routine laboratory procedures such as binding enzymes or coupling peptides to carrier proteins. These kits, mixture solutions, and collagen matrices fulfill a myriad of essential laboratory functions for developing relationships between proteins and other cellular components. The stimulating proteins offered have various amino acid arrangements and functions to fulfill any sample manipulation for testing purposes in any field.
Leupeptin
Supplier: VWR
A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin
Expand 2 Items
VWR Life Science Ribonuclease Inhibitor (RNase Inhibitor)
Supplier: VWR
RNase Inhibitor is used to inhibit the activity of RNases in reaction mixtures for cDNA synthesis and in vitro transcription, as well as for long term storage of valuable RNA samples
Expand 1 Items
Bovine Serum Albumin (BSA)
Supplier: VWR
Contains 0.1% sodium azide as preservative and strictly tested for RNase, DNase and Protease making it suitable for sensitive applications.
Expand 2 Items
Bovine Serum Albumin (BSA)
Supplier: VWR
Concentrated BSA solution (Albumin, Bovine, Fraction V – Cold Alcohol Isolation) in 0.85% sodium chloride and contains 0.1% sodium azide (as a preservative for stability).
Expand 1 Items
Aprotinin
Supplier: VWR
A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin
Expand 1 Items
Bovine Serum Albumin (BSA)
Supplier: VWR
Bovine serum albumin, powder, Fraction V. Nuclease and protease-free. Suitable for use in most molecular biology applications.
Expand 2 Items
Streptavidin
Supplier: VWR
A 60 kDa tetrameric protein that exhibits high binding affinity for biotin. Able to bind one molecule of biotin with each subunit, streptavidin is useful in affinity chromatography, ELISA, immunohistochemistry and Western Blotting.
Expand 2 Items
Bovine Serum Albumin (BSA)
Supplier: VWR
Crystalline. Prepared by a cold alcohol isolation procedure. This salt-free preparation is the purest material ideal for sensitive biochemical and diagnostic assays. DNase- and RNase-free, virtually free of globulins and other interfering contaminants.
Expand 1 Items
J.T.Baker®, Chicken Native Albumin (from Egg)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS RN 9006-50-2. Powder, Purified. Packaged in a plastic container. 500g
Expand 2 Items
Protein Phosphatase 2 Substrate
Supplier: Enzo Life Sciences
This peptide sequence, RRApTVA, has been found to be an excellent substrate for both PP2A and PP2C phosphatases.
Expand 1 Items
Amara Substrate Peptide
Supplier: Enzo Life Sciences
AMPK and SIK kinase substrate
Expand 1 Items
SAMS peptide (phosphorylated),Biotin
Supplier: Enzo Life Sciences
AMPK substrate
Expand 1 Items
Cdk5 Substrate
Supplier: Enzo Life Sciences
This peptide, PKTPKKAKKL, is derived from histone H1 and is a substrate for CDK5.
Expand 1 Items
Histone H3 Dimethyl Lysine-4 Peptide
Supplier: Enzo Life Sciences
Demethylase substrate
Expand 1 Items
NBD peptide
Supplier: Enzo Life Sciences
For increased cell permeability the peptide was covalently linked to the 10 aa recognized by the TAT transporter.