Order Entry
Canada
ContactUsLinkComponent
Outdoor Labels for Moist, Frosty or Frozen Surfaces, Diversified Biotech
Outdoor Labels for Moist, Frosty or Frozen Surfaces, Diversified Biotech
Catalog # 10141-760
undefined
Outdoor Labels for Moist, Frosty or Frozen Surfaces, Diversified Biotech
Catalog # 10141-760
Supplier Number:  GRRL-1000

Specifications

  • For Use With:
    1.5 ml tubes
  • Color:
    White
  • Description:
    Wetgrip labels
  • Dimensions:
    33×13 mm (1.29×0.51")
  • Environmentally Preferable:
  • Cat. no.:
    10141-760
  • Supplier No.:
    GRRL-1000

Specifications

About this item

Grips to wet, frost-covered and frozen surfaces; temperature range –80 to100 °C.

  • Thin material is compatible with centrifuge rotors
  • Resistant to chemicals and solvents
  • Adheres to plastics, glass, metals and even paperboard boxes

Choose your format: laser sheets for printing many labels at once, direct thermal labels for printing a few labels at a time or labels on a roll for handwriting labels.