ContactUsLinkComponent
PMP Erlenmeyer Flasks with Closure
PMP Erlenmeyer Flasks with Closure
Catalog # 470205-834
PMP Erlenmeyer Flasks with Closure
Catalog # 470205-834
Supplier Number:  4109-0125
Specifications
  • Capacity:
    125 ml
  • Cat. no.:
    470205-834
  • Supplier No.:
    4109-0125
Specifications
About this item
Provide a Combination of Chemical Resistance, Strength and Transparency.
  • Constructed with sturdy polymethylpentene
  • Excellent of culture media preparation and storage
  • Cap fits tightly with screw closure