Order Entry
ContactUsLinkComponent
12594 results for Proteins and Peptides

You searched for: Proteins and Peptides

Proteins and Peptides

Proteins are used in routine laboratory procedures such as binding enzymes or coupling peptides to carrier proteins. These kits, mixture solutions, and collagen matrices fulfill a myriad of essential laboratory functions for developing relationships between proteins and other cellular components. The stimulating proteins offered have various amino acid arrangements and functions to fulfill any sample manipulation for testing purposes in any field.

Sort by
Leupeptin

Leupeptin

Supplier: VWR

A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin

Expand 2 Items
Loading...
Aprotinin

Aprotinin

Supplier: VWR

A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin

Expand 1 Items
Loading...
Streptavidin

Streptavidin

Supplier: VWR

A 60 kDa tetrameric protein that exhibits high binding affinity for biotin. Able to bind one molecule of biotin with each subunit, streptavidin is useful in affinity chromatography, ELISA, immunohistochemistry and Western Blotting.

Expand 2 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Contains 0.1% sodium azide as preservative and strictly tested for RNase, DNase and Protease making it suitable for sensitive applications.

Expand 2 Items
Loading...
Leupeptin

Leupeptin

Supplier: VWR

Product is animal-free

Expand 1 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Concentrated BSA solution (Albumin, Bovine, Fraction V – Cold Alcohol Isolation) in 0.85% sodium chloride and contains 0.1% sodium azide (as a preservative for stability).

Expand 1 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Bovine serum albumin, powder, Fraction V. Nuclease and protease-free. Suitable for use in most molecular biology applications.

Expand 2 Items
Loading...
Casein

Casein

Supplier: VWR

High purity grade milk protein powder

Expand 1 Items
Loading...
J.T.Baker®, Chicken Native Albumin (from Egg)

J.T.Baker®, Chicken Native Albumin (from Egg)

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

CAS RN 9006-50-2. Powder, Purified. Packaged in a plastic container. 500g

Expand 2 Items
Loading...
Gelatin

Gelatin

Supplier: VWR

Gelatin

Expand 2 Items
Loading...
Bovine Serum Albumin (BSA)
Expand 2 Items
Loading...
Aprotinin

Aprotinin

Supplier: VWR

Product is animal-free

Expand 1 Items
Loading...
VWR Life Science Ribonuclease Inhibitor (RNase Inhibitor)

VWR Life Science Ribonuclease Inhibitor (RNase Inhibitor)

Supplier: VWR

RNase Inhibitor is used to inhibit the activity of RNases in reaction mixtures for cDNA synthesis and in vitro transcription, as well as for long term storage of valuable RNA samples

Expand 1 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Crystalline. Prepared by a cold alcohol isolation procedure. This salt-free preparation is the purest material ideal for sensitive biochemical and diagnostic assays. DNase- and RNase-free, virtually free of globulins and other interfering contaminants.

Expand 1 Items
Loading...
Protein Phosphatase 2 Substrate

Protein Phosphatase 2 Substrate

Supplier: Enzo Life Sciences

This peptide sequence, RRApTVA, has been found to be an excellent substrate for both PP2A and PP2C phosphatases.

Expand 1 Items
Loading...
SAMS peptide

SAMS peptide

Supplier: Enzo Life Sciences

AMPK substrate

Expand 1 Items
Loading...
PDKtide, Biotin

PDKtide, Biotin

Supplier: Enzo Life Sciences

Protein kinase substrate

Expand 1 Items
Loading...
Amara Substrate Peptide

Amara Substrate Peptide

Supplier: Enzo Life Sciences

AMPK and SIK kinase substrate

Expand 1 Items
Loading...
SAMS peptide (phosphorylated),Biotin

SAMS peptide (phosphorylated),Biotin

Supplier: Enzo Life Sciences

AMPK substrate

Expand 1 Items
Loading...
Human Recombinant Ubiquitin

Human Recombinant Ubiquitin

Supplier: Enzo Life Sciences

Expand 1 Items
Loading...
Cdk5 Substrate

Cdk5 Substrate

Supplier: Enzo Life Sciences

This peptide, PKTPKKAKKL, is derived from histone H1 and is a substrate for CDK5.

Expand 1 Items
Loading...
Histone H3 Dimethyl Lysine-4 Peptide

Histone H3 Dimethyl Lysine-4 Peptide

Supplier: Enzo Life Sciences

Demethylase substrate

Expand 1 Items
Loading...
E-64-C

E-64-C

Supplier: Enzo Life Sciences

Calpain and cathepsin inhibitor

Expand 2 Items
Loading...
GP antagonist-2A

GP antagonist-2A

Supplier: Enzo Life Sciences

G protein antagonist

Expand 1 Items
Loading...
Human Humanin

Human Humanin

Supplier: Enzo Life Sciences

Bax inhibitor

Expand 2 Items
Loading...

CinnGEL 2-methylester

Supplier: Enzo Life Sciences

PTP1B inhibitor

Expand 1 Items
Loading...
Z-LEHD-Fmk

Z-LEHD-Fmk

Supplier: Enzo Life Sciences

Caspase-9 inhibitor

Expand 2 Items
Loading...
AC-DMQD-AMC

AC-DMQD-AMC

Supplier: Enzo Life Sciences

Caspase-3 substrate

Expand 1 Items
Loading...
NBD peptide

NBD peptide

Supplier: Enzo Life Sciences

For increased cell permeability the peptide was covalently linked to the 10 aa recognized by the TAT transporter.

Expand 1 Items
Loading...
Leupeptin

Leupeptin

Supplier: Enzo Life Sciences

Cathepsin B inhibitor

Expand 3 Items
Loading...
Sort by
Recommended for You