Order Entry
Canada
ContactUsLinkComponent
NHS-Activated Sepharose™ 4 Fast Flow Affinity Coupling Media, Cytiva
NHS-Activated Sepharose™ 4 Fast Flow Affinity Coupling Media, Cytiva
Catalog # CA95017-030
Supplier:  Cytiva
CAS Number:  
undefined
NHS-Activated Sepharose™ 4 Fast Flow Affinity Coupling Media, Cytiva
Catalog # CA95017-030
Supplier:  Cytiva
Supplier Number:  17-0906-01
CAS Number:  

Specifications

  • Particle Size:
    90 µm
  • Description:
    NHS-Activated Sepharose™ 4 Fast Flow
  • Size:
    25 mL
  • pH range:
    3 - 13
  • Matrix:
    4% cross-linked agarose
  • Ligand:
    N-hydroxysuccinimide (NHS) activated
  • Storage temperature:
    2 - 8 °C
  • Flow rate:
    150 - 250 cm/h
  • Cat. no.:
    CA95017-030
  • Type:
    Liquid
  • Supplier No.:
    17-0906-01

Specifications

About this item

NHS-activated Sepharose™ 4 Fast Flow is composed of cross-linked 4% agarose beads, NHS activated for coupling amino-containing proteins and peptides in process-scale applications. NHS (N-hydroxysuccinimide) coupling forms a chemically stable amide bond with ligands containing primary amino groups. The resin provides a spacer arm to the coupled ligand and suitable for immobilising smaller protein and peptide ligands.

  • For affinity chromatography
  • NHS pre-activated medium for coupling of small amino-containing proteins and peptides in process-scale applications
  • NHS coupling method results in chemically stable ligand attachment
  • BioProcess medium supported for industrial applications
  • High level of pre-activation gives a high degree of substitution of the selected ligand
  • Stable in commonly used aqueous buffers provided that the ligand withstand the pH or the chemical environment

NHS-activated Sepharose 4 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, NHS-activated Sepharose 4 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support. This resin provides a spacer arm to the coupled ligand and is therefore particularly suitable for immobilising smaller protein and peptide ligands.

NHS (N-hydroxysuccinimide) coupling forms a chemically stable amide bond with ligands containing primary amino groups.