Affinity purification media, GammaBind™ Plus Sepharose™
Supplier: Cytiva
Certificates
About this item
GammaBind Plus Sepharose is GammaBind G, Type 3, covalently immobilized to Sepharose CL-6B by malimide linkage.
- For affinity chromatography
- Recombinant Protein G type 3 coupled to Sepharose CL-6B for enhanced binding to rat and mouse IgG subclasses at laboratory scale
- Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
- Maleimide activation coupling method
- Yields up to 18 mg/mL mouse IgG
- Stable in all commonly used aqueous buffers
GammaBind G, Type 3, a recombinant form of streptococcal protein G, binds to the Fc region of IgG from a variety of mammalian species. GammaBind Plus Sepharose may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium. Since only the Fc region is involved in binding, the Fab region is still available for binding antigen.
Specifications
- pH range:3 - 9
- Matrix:Cross-linked agarose, 6%
- Ligand:Recombinant GammaBind G type 3 lacking albumin-binding region
- Storage temperature:2 - 8 °C
- Binding capacity:> 35 mg human IgG/mL
- Flow rate:<130 cm/h
Specifications
Related Information
Certificates
Search by Lot Number17-0886-01_42 IN 17-0886-01_42 IN (1) 17-0886-04_42 IN 17-0886-04_42 IN (1) 17088601_INSTRUCTIONS
Manual
More
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together