EAH Sepharose™ 4B Affinity Coupling Media, Cytiva
Catalog # CA95016-888L
Supplier: Cytiva
CAS Number:
Specifications
- Particle Size:90 µm
- Description:EAH Sepharose™ 4B
- Size:50 mL
- Matrix:Agarose, 4%
- Storage temperature:4 - 8 °C
- Flow rate:<75 cm/h
- Cat. no.:CA95016-888L
- Type:Liquid
- Supplier No.:17-0569-01
Specifications
About this item
EAH Sepharose™ 4B allows for the stable carbodiimide-based coupling of carboxyl groups to Sepharose® 4B via an 11-atom hydrophilic spacer arm that permits extremely stable coupling.
Stable carbodiimide-based coupling of carboxyl groups to 4B via an 11-atom hydrophilic spacer arm permits very stable coupling. Stable to all commonly used aqueous Sepharose buffers.