Peristaltic Pump, Cytiva
Catalog #: CA10500-518
Supplier: Cytiva
Specifications
- Description:Peristaltic Pump
- Cat. no.:CA10500-518
- Supplier No.:29-0239-92
Specifications
About this item
This single-channel peristaltic pump consists of a four-roller pump head that delivers accurate, low pulsation flow rates to enable reproducible isocratic or gradient elution.
The pump provides flow rates up to 5 ml/min.
Customers Who Bought This Also Bought