ContactUsLinkComponent
Test Papers, pH, Ion, Litmus, Advantec MFS
  102939-570
undefined
Test Papers, pH, Ion, Litmus, Advantec MFS
  102939-570
 :  UNIV-B
  • Description:
    UNIV (Universal)
  • Cat. no.:
    102939-570
  • Packaging:
    Booklet
  • Measurement Range:
    pH 1-11
  • Supplier No.:
    07010120
  • Paper Type:
    pH
  • Quantity Per Box/Pack/Carton:
    10
pH Test Papers–Various types of indicator papers cover a wide pH range to allow for good accuracy of determining pH value of a solution
Ion Test Papers–Detects concentration of specified ion in solution. Two-point indicator strip confirms within pH range and ion concentration by color comparison.
Litmus Paper–General detection for whether solution is acidic (Blue) or alkaline (Red).
Dip the indicator test strip in solution and compare to the color chart on the container. For Litmus paper a color change is confirmation.
High sensitivity and sharp color change. Narrow pH unit graduation: 0.2-0.4 (UNIV/ WR: 1.0-2.0 units; Congo Red: color change to red indicates within specified range only).
 : Roll is 9mmx6M, except UNIV roll is 9mmx5.5M; Booklet: 7x70mm (20 leaves/booklet, 10 booklets/pk.). Strip: 5x85mm.