Corning® Gosselin™ Storage Bottles, Square, HDPE, Corning
Supplier: Corning
Certificates
About this item
Space saving bottles made of HDPE, translucent with HDPE or LDPE screw cap.
- Good chemical resistance, good temperature resistance
- Sterile oder packed under aseptic conditions
- Excellent shock resistance
Ideal for liquid, solid and powder storage. Good level of tightness according to the closure system used and application.
Related Information
Certificates
Search by Lot Number10799-438_750295 10799-438_770993
Conformity declaration
Corning Gosselin Square HDPE Storage Bottles.pdf Corning Plastic-Glass-Storage-Bottles Product-Selection-Guide Corning_Plastic_Storage_Bottles_Product_Selection_Guide.pdf Corning® Shopping list for returning to the bench F&BFOCUS Volume 5 Issue 1w
Product Literature
Corning New Lab Startup Program
Promotion Material
Customers Who Bought This Also Bought