Order Entry
Canada
ContactUsLinkComponent
Micro-Perfusion Pump Mounting Rod Clamp, Bioptechs Inc.®
Micro-Perfusion Pump Mounting Rod Clamp, Bioptechs Inc.®
Catalog # 10199-778
Supplier:  BIOPTECHS INC.
undefined
Micro-Perfusion Pump Mounting Rod Clamp, Bioptechs Inc.®
Catalog # 10199-778
Supplier:  BIOPTECHS INC.
Supplier Number:  16181303

Specifications

  • Description:
    Micro-Perfusion Pump Mounting Rod Clamp
  • Cat. no.:
    10199-778
  • Supplier No.:
    16181303

Specifications

About this item

A polymer screw-on clamping bracket to support the Bioptechs Micro-Perfusion Pump onto conventional 1.27cm (0.5") breadboard support post.

Certifications: CE certified.