ContactUsLinkComponent
FCS2® Open Mode Adapter, Bioptechs Inc.®
FCS2® Open Mode Adapter, Bioptechs Inc.®
Catalog # 10200-022
Supplier:  BIOPTECHS INC.
undefined
FCS2® Open Mode Adapter, Bioptechs Inc.®
Catalog # 10200-022
Supplier:  BIOPTECHS INC.
Supplier Number:  06031921513
Specifications
  • Description:
    FCS2® Open Mode Adapter
  • Cat. no.:
    10200-022
  • Supplier No.:
    06031921513
Specifications
About this item
The white, top portion of a FCS2® without perfusion ports, which enables the FCS2® to be assembled without the microaqueduct slide
This enables sealing of the 40 mm coverslip to the base of the chamber. Oftentimes used for microinjection.
Certifications: CE certified.