Rat Recombinant IL-6 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Interleukin 6 (IL-6) is a cytokine that is expressed by T cells, macrophages, and muscle cells. IL-6 acts to stimulate an immune response upon infection or trauma. IL-6 has both pro-inflammatory and anti-inflammatory functions, is capable of promoting fever, and signals through the cell-surface type I cytokine receptor complex containing the IL-6Ra and gp130 chains.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: B9 cell proliferation, ED50 ≤2 ng/ml, bioactivity ≥5.0×10⁵ units/mg
- Gene ID:P20607
- Reconstitution instructions:Sterile 10 mM HCl at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:IFN-β2|HSF|BSF-2|B-cell differentiation factor|MGI-2
- UniProtKB:P20607
- Protein/peptide name:IL-6
- Purity:≥90%
- Molecular weight:21.9 kDa
- Sequence:MFPTSQVRRG DFTEDTTHNR PVYTTSQVGG LITYVLREIL EMRKELCNGN SDCMNSDDAL SENNLKLPEI QRNDGCFQTG YNQEICLLKI CSGLLEFRFY LEFVKNNLQD NKKDKARVIQ SNTETLVHIF KQEIKDSYKI VLPTPTSNAL LMEKLESQKE WLRTKTIQLI LKALEEFLKV TMRSTRQT
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA) + 2:1 mannitol to protein
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Related Information
Literature reference - PubMed ID 22763408
Additional Product Details
Shenandoah_Biotechnology_Product_Catalog
Product Literature
Frequently Bought Together