Order Entry
Canada
ContactUsLinkComponent
 
Rat Recombinant TNFalpha (from E. coli)
 
undefined
Rat Recombinant TNFalpha (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED50 ≤0.05 ng/ml, Bioactivity ≥2.0×10⁷ units/mg
  • Gene ID:
    P16599
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    Cytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|Cachectin
  • Protein/peptide name:
    TNFalpha
  • Purity:
    ≥95%
  • Molecular weight:
    17.3 kDa
  • Sequence:
    MLRSSSQNSS DKPVAHVVAN HQAEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLIY SQVLFKGQGC PDYVLLTHTV SRFATSYQEK VSLLSAIKSP CPKDTPEGAE LKPWYEPMYL GGVSQLEKGD LLSAEVNLPK YLDITESGQV YFGVIAL
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together