Order Entry
Canada
ContactUsLinkComponent
 
Rat Recombinant IL-17A (from E. coli)
 
undefined
Rat Recombinant IL-17A (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q61453
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    IL-17|CTLA-8
  • Protein/peptide name:
    IL-17A
  • Purity:
    ≥95%
  • Molecular weight:
    15.1/30.3 kDa
  • Sequence:
    MAVLIPQSSV CPNAEANNFL QNVKVNLKVL NSLSSKASSR RPSDYLNRST SPWTLSRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPEKCPF TFRVEKMLVG VGCTCVSSIV RHAS
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium citrate, pH 3.0
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

 

Frequently Bought Together