Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Mouse Recombinant RELM-B (from E. coli)

Mouse Recombinant RELM-B (from <i>E. coli</i>)
 
Mouse Recombinant RELM-B (from <i>E. coli</i>)
Mouse Recombinant RELM-B (from E. coli)

 

 

Interleukin-19 (IL-19) is a member of the interleukin 10 (IL-10) cytokine family and is produced by B cells and monocytes. IL-19 binds the interleukin 20 receptor complex (IL-20R) to activate STAT3 signaling. IL-19 induces interleukin 6 (IL-6) and tumor necrosis factor alpha (TNF-α) expression in monocytes, and promotes type 2 T helper (Th2) cell-mediated immune responses. IL-19 production is upregulated in resting monocytes following granulocyte-macrophage colony-stimulating factor (GM-CSF) or lipopolysaccharide (LPS) stimulation.

  • High quality
  • Low endotoxin
  • Affordable

Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.

 : For research use only.

 

 

 

  • Catalog No:
  • 10788-342
  • 10788-340
  • 10788-336
  • 10822-350
  • 10788-338
  • Protein/peptide name:
  • RELM-B
  • RELM-B
  • RELM-B
  • RELM-B
  • RELM-B
  • Protein synonyms:
  • FIZZ2
  • FIZZ2
  • FIZZ2
  • FIZZ2
  • FIZZ2
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • Q99P86
  • Q99P86
  • Q99P86
  • Q99P86
  • Q99P86
  • Sequence:
  • MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
  • MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
  • MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
  • MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
  • MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • UniProtKB:
  • Q99P86
  • Q99P86
  • Q99P86
  • Q99P86
  • Q99P86
  • Biological activity:
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • Molecular weight:
  • 8.9/17.8 kDa
  • 8.9/17.8 kDa
  • 8.9/17.8 kDa
  • 8.9/17.8 kDa
  • 8.9/17.8 kDa
  • Reconstitution instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage conditions:
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT

 

Product Family Options

 

  • Pack type
    Size
    Origin
     
     
  • Vial
    5 µg
    US
       
       
    Each (5µG)
    Each (5µG) - $146.49
    $146.49
    Each (5µG)
       $146.49
     

     : 200-69-5UG
     
     
    Specifications
    Cat. No.
    10788-342
    Protein/peptide name
    RELM-B
    Protein synonyms
    FIZZ2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q99P86
    Sequence
    MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q99P86
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8.9/17.8 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    25 µg
    US
       
       
    Each (25µG)
    Each (25µG) - $366.60
    $366.60
    Each (25µG)
       $366.60
     

     : 200-69-25UG
     
     
    Specifications
    Cat. No.
    10788-340
    Protein/peptide name
    RELM-B
    Protein synonyms
    FIZZ2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q99P86
    Sequence
    MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q99P86
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8.9/17.8 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    100 µg
    US
       
       
    Each (100µG)
    Each (100µG) - $915.69
    $915.69
    Each (100µG)
       $915.69
     

     : 200-69-100UG
     
     
    Specifications
    Cat. No.
    10788-336
    Protein/peptide name
    RELM-B
    Protein synonyms
    FIZZ2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q99P86
    Sequence
    MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q99P86
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8.9/17.8 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    500 µg
    US
       
       
    Each (500µG)
    Each (500µG) - $3,835.03
    $3,835.03
    Each (500µG)
       $3,835.03
     

     : 200-69-500UG
     
     
     
    Specifications
    Cat. No.
    10822-350
    Protein/peptide name
    RELM-B
    Protein synonyms
    FIZZ2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q99P86
    Sequence
    MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q99P86
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8.9/17.8 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    1 mg
    US
       
       
    Each (1mg)
    Each (1mg) - $6,598.86
    $6,598.86
    Each (1mg)
       $6,598.86
     

     : 200-69-1MG
     
     
    Specifications
    Cat. No.
    10788-338
    Protein/peptide name
    RELM-B
    Protein synonyms
    FIZZ2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q99P86
    Sequence
    MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q99P86
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8.9/17.8 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.