Rat Recombinant FGF-9 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Fibroblast growth factor 9 (FGF-9) is a mitogen and survival factor for nerve and mesenchymal cells. FGF-9 functions as an autocrine and paracrine factor to support the growth and survival of motor neurons and prostate tissue. FGF-9 expression in the gonad is also necessary for sex determination.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P36364
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Heparin-binding growth factor-9|Glial activating factor|GAF|HBGF-9
- Protein/peptide name:FGF-9
- Purity:≥95%
- Molecular weight:23.3 kDa
- Sequence:MPLGEVGSYF GVQDAVPFGN VPVLPVDSPV LLSDHLGQSE AGGLPRGPAV TDLDHLKGIL RRRQLYCRTG FHLEIFPNGT IQGTRKDHSR FGILEFISIA VGLVSIRGVD SGLYLGMNEK GELYGSEKLT QECVFREQFE ENWYNTYSSN LYKHVDTGRR YYVALNKDGT PREGTRTKRH QKFTHFLPRP VDPDKVPELY KDILSQS
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, 75 mM Ammonium sulfate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together