Mouse Recombinant IP-10 (from E. coli)
Supplier: Fujifilm Irvine Scientific
Certificates
About this item
Macrophage inflammatory protein-1 alpha (MIP-1 α), also known as CCL3, is a cytokine produced by macrophages. MIP-1 α binds the chemokine receptors CCR1, CCR4, and CCR5 to induce inflammatory responses, including the recruitment of granulocytes and neutrophil superoxide production. The MIP-1 α and MIP-1 β heterodimer exhibits antiviral activity against the human immunodeficiency virus 1 (HIV-1).
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P17515
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CXCL10|CRG-2
- UniProtKB:P17515
- Protein/peptide name:IP-10
- Purity:≥95%
- Molecular weight:8.7 kDa
- Sequence:IPLARTVRCN CIHIDDGPVR MRAIGKLEII PASLSCPRVE IIATMKKNDE QRCLNPESKT IKNLMKAFSQ KRSKRAP
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Related Information
Literature reference - PubMed ID 20405477 Literature reference - PubMed ID 25986989
Additional Product Details
Shenandoah_Biotechnology_Product_Catalog
Product Literature
Frequently Bought Together