Human Recombinant FGF-10 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Fibroblast growth factor 10 (FGF-10) is a growth factor that is important during embryonic development, especially during lung, limb, brain, heart, and kidney morphogenesis. FGF-10 is expressed in mesenchymal cells and facilitates epithelial-mesenchymal signaling through binding the epithelially expressed FGF receptor 2b (FGFR2b). FGF-10 also functions as a mitogen for keratinizing epidermal cells, and induces the migration and invasion of cancer cells.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5.0×10³ units/mg
- Gene ID:O15520
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Keratinocyte growth factor 2
- Protein/peptide name:FGF-10
- Purity:≥95%
- Molecular weight:19.3 kDa
- Sequence:MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEIGVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Related Information
Frequently Bought Together