Order Entry
Canada
ContactUsLinkComponent
Anti-IgG Rat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgG Rat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalogue # RL611-502-122
undefined
Anti-IgG Rat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalogue # RL611-502-122
Supplier Number:  611-502-122
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Rabbit
  • Size:
    1 mg
  • Epitope:
    Heavy and Light chain
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
  • Storage temperature:
    –20 °C
  • Concentration:
    1.0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ambient
  • Immunogen:
    Rabbit IgG whole molecule
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. no.:
    RL611-502-122
  • Supplier No.:
    611-502-122

Spécifications

À propos de cet article

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and Light chain
Host: Rat
Isotype:
Reactivity: Rabbit