Anti-ALS2CL Rabbit Polyclonal Antibody
Catalog # 89517-892
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:ALS2CL
- Antigen symbol:ALS2CL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:400 µL
- Epitope:Center
- Gene ID:259173
- Antigen synonyms:RN49018
- Amino acid number:349 to 377
- Storage buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular weight:107.748 kDa
- Sequence:RGRPHGKGTLKWPDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping temperature:4 °C
- Immunogen:This ALS2CL antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 349-377 amino acids from the Central region of human ALS2CL.
- Purification:Protein A & Peptide affinity purification
- Cat. no.:89517-892
- Supplier No.:AP17574C
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: ALS2CL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human