About this item
FITC anti-human IL-4 [MP4-25D2]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Swine (Pig, Porcine), Rhesus; Apps: ICFC; Size: 50 μg
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.
Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Human;Cross-Reactivity: Swine (Pig;Porcine);Rhesus
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA500806-BL
- CA500807-BL
- Antigen symbol:
- IL-4
- IL-4
- Antigen synonyms:
- IL4|MCGF-2|IaIF|HCGF|TCGF-2|MFF|BSF-1|Interleukin-4
- IL4|MCGF-2|IaIF|HCGF|TCGF-2|MFF|BSF-1|Interleukin-4
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- MP4-25D2
- MP4-25D2
- Reactivity:
- Pig, Rhesus monkey, Human
- Pig, Rhesus monkey, Human
- Host:
- Rat
- Rat
- Gene ID:
- 3565
- 3565
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- CHO-expressed, recombinant human IL-4
- CHO-expressed, recombinant human IL-4
- Form:
- test size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- test size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- Concentration:
- 0.50 mg/mL
- Storage temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT