Order Entry
Canada
ContactUsLinkComponent
Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # RL702-400-002
Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # RL702-400-002
Supplier Number:  702-400-002

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Cat
  • Size:
    500 µL
  • Epitope:
    Heavy and Light chain
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Antigen synonyms:
    F(ab')2 Anti-Cat IgG (H&L)
  • Antibody fragment:
    F(ab')2
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Cat IgG whole molecule
  • Tested applications:
    Fluoresence ELISA, Fluoresence Western Blot
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. no.:
    RL702-400-002
  • Supplier No.:
    702-400-002

Specifications

About this item

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Cat