Order Entry
Canada
ContactUsLinkComponent
Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: RM4-5]
Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: RM4-5]
Supplier:  Biolegend
Certificates
Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: RM4-5]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • CA10758-842
  • CA100544-BL
  • CA100543-BL
  • Antigen symbol:
  • CD4
  • CD4
  • CD4
  • Antigen synonyms:
  • L3T4|T4
  • L3T4|T4
  • L3T4|T4
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Brilliant Violet® 421
  • Brilliant Violet® 421
  • Brilliant Violet® 421
  • Clone:
  • RM4-5
  • RM4-5
  • RM4-5
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 12504
  • 12504
  • 12504
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Immunogen:
  • BALB/c mouse thymocytes
  • BALB/c mouse thymocytes
  • BALB/c mouse thymocytes
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
  • Concentration:
  • 0.20 mg/mL
  • Storage temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
  • Ambient RT
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together