Anti-SSRP1 Mouse Monoclonal Antibody [clone: 10D1]
Supplier: Biolegend
Certificates
About this item
Purified anti-SSRP1 [10D1]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: WB, IF, ChIP, IP; Size: 100 μg
Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: SSRP1
Clonality: Monoclonal
Clone: 10D1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Specifications
- Catalog No:
- CA609701-BL
- CA609702-BL
- Antigen symbol:
- SSRP1
- SSRP1
- Antigen synonyms:
- Facilitates Chromatin Transcription 80 kD subunit|Structure specific recognition protein 1 (SSRP1)|Recombination signal sequence recognition protein
- Facilitates Chromatin Transcription 80 kD subunit|Structure specific recognition protein 1 (SSRP1)|Recombination signal sequence recognition protein
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 10D1
- 10D1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 6749
- 6749
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Immunogen:
- Recombinant (partial) (308-524aa)
- Recombinant (partial) (308-524aa)
- Form:
- This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide at 0.5 mg/ml.
- This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide at 0.5 mg/ml.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage temperature:
- Upon receipt, store undiluted between 2 °C and 8 °C.
- Upon receipt, store undiluted between 2 °C and 8 °C.
- Shipping temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together