Anti-UBE2D2 Rabbit Polyclonal Antibody
Catalog # 89520-962
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating Enzyme E2D2
- Antigen symbol:UBE2D2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:7322
- Antigen synonyms:UBCH4|E2(17)KB2|UBCH5B|PUBC1|UBC4/5|UBC4
- Amino acid number:112 to 140
- Storage buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular weight:16.735 kDa
- Sequence:YKTDREKYNRIAREC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping temperature:4 °C
- Immunogen:This UBE2D2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 112-140 amino acids from the C-terminal region of human UBE2D2.
- Purification:Protein A & Peptide affinity purification
- Cat. no.:89520-962
- Supplier No.:AP14752B
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: UBE2D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human