Order Entry
Canada
ContactUsLinkComponent
Anti-CD274 (B7-H1, PD-L1) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 29E.2A3]
Anti-CD274 (B7-H1, PD-L1) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 29E.2A3]
Supplier:  Biolegend
Certificates
Anti-CD274 (B7-H1, PD-L1) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 29E.2A3]
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • CA329706-BL
  • CA329705-BL
  • Antigen symbol:
  • CD274 (B7-H1, PD-L1)
  • CD274 (B7-H1, PD-L1)
  • Antigen synonyms:
  • PDL1|Programmed cell death ligand 1 (PD-L1)|29E2A3|B7 homolog 1 (B7-H1) B7H1|CD274|Immune checkpoints
  • PDL1|Programmed cell death ligand 1 (PD-L1)|29E2A3|B7 homolog 1 (B7-H1) B7H1|CD274|Immune checkpoints
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 29E.2A3
  • 29E.2A3
  • Reactivity:
  • Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
  • Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 29126
  • 29126
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Full length human PD-L1
  • Full length human PD-L1
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • Storage temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information

Frequently Bought Together