Order Entry
Canada
ContactUsLinkComponent
Anti-Ly-6A/E (Sca-1) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: D7]
Anti-Ly-6A/E (Sca-1) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: D7]
Supplier:  Biolegend
Certificates
Anti-Ly-6A/E (Sca-1) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: D7]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • CA108119-BL
  • CA108120-BL
  • Antigen symbol:
  • Ly-6A/E (Sca-1)
  • Ly-6A/E (Sca-1)
  • Antigen synonyms:
  • Sca-1|Ly-6A/E
  • Sca-1|Ly-6A/E
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PB (Pacific Blue)
  • PB (Pacific Blue)
  • Clone:
  • D7
  • D7
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 110454
  • 110454
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • IL-2-dependent mouse T-cell line (CTL-L)
  • IL-2-dependent mouse T-cell line (CTL-L)
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
  • The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
  • Concentration:
  • 0.50 mg/mL
  • 0.50 mg/mL
  • Storage temperature:
  • The Ly-6A/E antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The Ly-6A/E antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information

Frequently Bought Together