About this item
Pacific Blue™ anti-human CD45RA [HI100]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test size, the suggested use of this reagent for immunofluorescent staining is 20 μl per million cells in 100 μl volume.
For μg sizes, the suggested use of this reagent for immunofluorescent staining is 0.5 μg per million cells in 100 μl volume.
It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: HI100
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Specifications
- Catalog No:
- CA304117-BL
- CA304118-BL
- CA304123-BL
- Antigen symbol:
- CD45RA
- CD45RA
- CD45RA
- Antigen synonyms:
- CD45R|B220|CD45|PTPRC|GP180|T200|L-CA|LY5|LCA
- CD45R|B220|CD45|PTPRC|GP180|T200|L-CA|LY5|LCA
- CD45R|B220|CD45|PTPRC|GP180|T200|L-CA|LY5|LCA
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- PB (Pacific Blue)
- PB (Pacific Blue)
- PB (Pacific Blue)
- Clone:
- HI100
- HI100
- HI100
- Reactivity:
- Chimpanzee, Human
- Chimpanzee, Human
- Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 5788
- 5788
- 5788
- Isotype:
- IgG2b kappa
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Yes
- Form:
- test size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- test size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- test size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
- The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
- The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage temperature:
- The CD45RA antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The CD45RA antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The CD45RA antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT
- Ambient RT