Order Entry
ContactUsLinkComponent
446 results for Vials

You searched for: Vials

Vials

Vials are available in all capacities to meet any collection, storage, or analysis need. Tailor vessels for specific media with various translucent manufacturing and different sterility options for precise test results. Choose from the extensive selection featuring autoclavable capabilities for temperature testing. Styles come without or with caps that may include liners or septas for easy specimen handling. Correctly identify specimen immediately with the write-on labels to eliminate testing errors.

Sort by
VWR® 11 mm Snap Ring Vials, Kits, Caps, and Inserts, Wide Opening

VWR® 11 mm Snap Ring Vials, Kits, Caps, and Inserts, Wide Opening

Supplier: VWR International

11 mm snap ring vials can be used with snap caps or aluminum crimp seal closures.

Expand 7 Items
Loading...
VWR® Shell Vial and Plug Cap

VWR® Shell Vial and Plug Cap

Supplier: VWR International

Designed for use in Alcott®, Waters®, Gilson®, or Shimadzu® autosamplers, the clear snap-plug closures of these shell vials can be pierced with standard autosampler syringes without vacuum formation.

Expand 1 Items
Loading...
VWR® Scintillation Vials with Screw Cap, Borosilicate Glass

VWR® Scintillation Vials with Screw Cap, Borosilicate Glass

Supplier: VWR International

VWR® scintillation vials are produced from borosilicate 3.3 glass. They are compatible with all scintillation counters.

Expand 8 Items
Loading...
VWR® Drosophila Vials, Narrow and Wide

VWR® Drosophila Vials, Narrow and Wide

Supplier: VWR International

Variety of sizes and materials for all applications.

Expand 12 Items
Loading...
VWR® 18 mm Screw-Thread Headspace Vials and Caps

VWR® 18 mm Screw-Thread Headspace Vials and Caps

Supplier: VWR International

Screw-thread magnetic headspace vials offer tool-free convenience of a screw-thread vial

Expand 2 Items
Loading...
VWR® Vials, Borosilicate Glass, with Phenolic Screw Cap

VWR® Vials, Borosilicate Glass, with Phenolic Screw Cap

Supplier: VWR International

These general-purpose vials are available in either a short form or capsule style.

Expand 12 Items
Loading...
VWR® 11 mm Wide Opening Crimp-Top Vials

VWR® 11 mm Wide Opening Crimp-Top Vials

Supplier: VWR International

Wide opening vials feature 40% larger neck opening than standard opening vials, for easier access and reduced needle damage.

Expand 1 Items
Loading...
VWR® Screw-Thread Vials 8-425, Caps and Inserts with Standard Opening

VWR® Screw-Thread Vials 8-425, Caps and Inserts with Standard Opening

Supplier: VWR International

8-425 autosampler screw thread vials feature a standard opening design compatible with 5 mm inserts.

Expand 6 Items
Loading...
VWR® 4 ml Screw-Thread Vials, Kits, and Caps

VWR® 4 ml Screw-Thread Vials, Kits, and Caps

Supplier: VWR International

4 ml screw thread vials are designed for use with Waters® WISP® (48-position) and Shimadzu™ autosamplers.

Expand 6 Items
Loading...
VWR® Vials, PTFE

VWR® Vials, PTFE

Supplier: VWR International

PTFE vials are ideal for storing aggressive or valuable materials.

Expand 2 Items
Loading...
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening

VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening

Supplier: VWR International

9 mm autosampler screw thread vials feature a wide opening design for compatibility with all major autosampler brands and robotics.

Expand 28 Items
Loading...
VWR® Inserts for Standard Opening Vials

VWR® Inserts for Standard Opening Vials

Supplier: VWR International

VWR Insert, Standard Flat-Bottom Glass, for Standard Opening Vials, 5x31 mm, Clear, Capacity: 250ul

Expand 1 Items
Loading...
VWR® 10 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening

VWR® 10 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening

Supplier: VWR International

10-425 autosampler screw thread vials feature a wide opening compatible with 6 mm inserts.

Expand 5 Items
Loading...

VWR® Screw Thread Vial with Cap

Supplier: VWR International

Made of type 1 clear borosilicate glass, these screw thread vials are designed with a continuous thread 'CT' finish, for a positive seal in a variety of uses including laboratory or manufacturer's use.

Expand 2 Items
Loading...
VWR® TraceClean® Vials with Chemical Preservative

VWR® TraceClean® Vials with Chemical Preservative

Supplier: VWR International

Type I borosilicate glass vials contain a chemical preservative to maintain sample integrity until volatile organic analysis can be performed.

Expand 3 Items
Loading...
VWR® Cryogenic Vials, Internally or Externally Threaded

VWR® Cryogenic Vials, Internally or Externally Threaded

Supplier: VWR International

Designed for storing biological material, human or animal cells at temperatures as low as –196 °C, but should be used only in the gas phase of liquid nitrogen.

Expand 3 Items
Loading...
VWR® TraceClean® 20 ml Vials

VWR® TraceClean® 20 ml Vials

Supplier: VWR International

Type I clear borosilicate glass vials are for use in various analyses and in specimen, extract, and standards storage.

Expand 6 Items
Loading...

VWR® TraceClean® Low TOC Vials

Supplier: VWR International

These glass vial are used primarily for the collection and testing of aqueous samples for Total Organic Carbon (TOC). Secondary uses include containment of extracts. Please refer to the applicable certificate of analysis for certified analytes and corresponding reporting limits.

Expand 2 Items
Loading...
VWR® Scintillation Vials, Borosilicate Glass, with Screw Caps

VWR® Scintillation Vials, Borosilicate Glass, with Screw Caps

Supplier: VWR International

These scintillation vials fit all standard counting equipment for liquid scintillation systems.

Expand 7 Items
Loading...
VWR® Ion Chromatography (IC) PolyVials

VWR® Ion Chromatography (IC) PolyVials

Supplier: VWR International

VWR® Ion Chromatography (IC) vials are compatible with Dionex® and Metrohm-Peak® Autosamplers.

Expand 5 Items
Loading...

VWR® TraceClean® Vials, Clear

Supplier: VWR International

These TraceClean® glass jars are used for the collection of and testing of aqueous samples for Volatile Organic Analysis and Total Petroleum Hydrocarbons (TPH).

Expand 7 Items
Loading...
VWR® 20 mm Crimp Seal Headspace Vials

VWR® 20 mm Crimp Seal Headspace Vials

Supplier: VWR International

Clear 20 ml headspace vials designed to fit most headspace autosamplers.

Expand 2 Items
Loading...
VWR® Cryovials

VWR® Cryovials

Supplier: VWR International

Designed for freezing samples in all cryopreservation applications, these vials feature an internal thread with washer, external thread. The caps and vials are made from the same batch of materials to ensure equal dilation coefficient, resulting in optimum sealing characteristics.

Expand 26 Items
Loading...
VWR® Microwave Reaction Vials

VWR® Microwave Reaction Vials

Supplier: VWR International

Vials are manufactured from Type I, Class A (USP Type I) heavy-wall borosilicate glass and are designed to withstand pressures up to 300 psi (20 bar).

Expand 3 Items
Loading...
VWR® 11 mm Crimp Top Vials, Kits, and Inserts, Standard Opening

VWR® 11 mm Crimp Top Vials, Kits, and Inserts, Standard Opening

Supplier: VWR International

These 11 mm crimp top, standard opening vials have a maximum fill volume of 1.8 ml and are manufactured from high purity borosilicate glass.

Expand 5 Items
Loading...
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening

VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening

Supplier: VWR International

11 mm crimp top, wide neck opening design, allows easy filling, requires inserts with a diameter of 6 mm.

Expand 8 Items
Loading...
VWR® Serum Vials / Diagnostic Bottles, PETG

VWR® Serum Vials / Diagnostic Bottles, PETG

Supplier: VWR International

All VWR® sterile labware, media bottles, vials, and culture vials are manufactured with the remarkable Eastar PETG resin from Eastman.

Expand 1 Items
Loading...
VWR® Inserts for VWR® 11 mm Standard Opening Crimp-Top Vials

VWR® Inserts for VWR® 11 mm Standard Opening Crimp-Top Vials

Supplier: VWR International

5mm insert for use with standard opening aluminum seal vials. Case of 100.

Expand 1 Items
Loading...
VWR® Inserts for Wide Opening Vials

VWR® Inserts for Wide Opening Vials

Supplier: VWR International

VWR Insert, Standard Flat-Bottom Glass, for Wide mouth Vials, 6x31 mm, Clear, Capacity: 500ul

Expand 4 Items
Loading...
VWR® 9 mm Screw-Thread Vials

VWR® 9 mm Screw-Thread Vials

Supplier: VWR International

Autosampler screw thread vials feature a wide mouth design for compatibility with all major autosampler brands and robotics.

Expand 1 Items
Loading...
Sort by
Recommended for You