Order Entry
Canada
ContactUsLinkComponent
 
Anti-SNVsSgp1 Rabbit Polyclonal Antibody
  10752-144
 :  Prosci
Anti-SNVsSgp1 Rabbit Polyclonal Antibody
  10752-144
 :  Prosci
 :  7685_S
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody type:
    Primary
  • Antigen name:
    SNVsSgp1 nucleocapsid
  • Antigen symbol:
    SNVsSgp1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Size:
    0.02 mg
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    Q9E1J8
  • Antigen synonyms:
    SNVsSgp1
  • Storage buffer:
    Sin Nombre virus nucleocapsid antibody is supplied in PBS containing 0.02% sodium azide.
  • Molecular weight:
    N/A
  • Storage temperature:
    Sin Nombre Nucleocapsid antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    4 °C
  • Immunogen:
    Sin Nombre virus nucleocapsid antibody was raised against a 19 amino acid peptide near the center of the Sin Nombre virus nucleocapsid. The immunogen is located within amino acids 170 - 220 of Sin Nombre Virus Nucleocapsid.
  • Purification:
    Sin Nombre virus nucleocapsid antibody is affinity chromatography purified via peptide column.
  • Cat. no.:
    10752-144
  • Supplier No.:
    7685_S

 

 

Sin Nombre virus (SNV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS). The hantavirus nucleocapsid protein plays several roles in viral replication and assembly, and is the major antigen in humoral responses in humans and mice. Within the bunyviridae family, the nucleocapsid protein also functions as an RNA chaperone facilitating the formation of the stable genomic RNA “panhandle” and is thought to aid in the replication of bunyavirus RNA.

Type: Primary
Antigen: SNVsSgp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus