To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:uracil-DNA glycosylase
- Antigen symbol:UNG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:0.02 mg
- Epitope:near the N-Terminus
- Cross adsorption:No
- Form:Liquid
- Gene ID:P13051
- Antigen synonyms:UNG
- Storage buffer:UNG2 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 34 kDa Observed: 33 kDa
- Storage temperature:UNG2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:UNG2 antibody was raised against a 17 amino acid peptide near the amino terminus of human UNG2. The immunogen is located within the first 50 amino acids of UNG2.
- Purification:UNG2 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10751-972
- Supplier No.:7353_S
UNG2 Antibody: The human uracil-DNA glycosylase (UNG) gene encodes both mitochondrial (UNG1) and nuclear (UNG2) forms through differentially regulated promoters and alternative splicing. UNG2 is the major enzyme in the base excision repair pathway that removes uracil residues from DNA that arise through either misincorporation during replication or cytosine deamination. UNG2 can also be bound by the HIV-1 integrase and incorporated into the virion particle, suggesting that it is required to remove uracils from the viral genome. As the intrinsic antiviral protein APOBEC3G generates numerous uracils in the HIV genome during its replication, it may be that the UNG2 contributes to the APOBEC3G-mediated loss of infectivity by generating abasic sites in the viral genome.
Type: Primary
Antigen: UNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse