Anti-UBD Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
UBD, also designated ubiquitin D or diubiquitin, plays a role in antigen presentation, cytokine response and apoptosis. It is a 165 amino acid protein encoded in the major histocompatibility complex (MHC) that consists of two domains which share significant homology with ubiquitin. UBD is inducible by interferon gamma and TNF-alpha and regulates TNF-alpha-induced and LPS-mediated activation of the central mediator of innate immunity NF-kappa-B by promoting TNF-alpha-mediated proteasomal degradation of ubiquitinated-I-kappa-B-alpha. It may also regulate cell growth during dendritic cell or B cell activation and development.
Type: Primary
Antigen: UBD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
10749-230.pdf
Datasheet
Specifications
- Size:0.02 mg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:UBD antibody is affinity chromatography purified via peptide column.
- Antibody type:Primary
- Form:Liquid
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:UBD antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Host:Rabbit
- Concentration:1 mg/mL
- Cross adsorption:No
- Molecular weight:Predicted: 18 kDa Observed: 17 kDa
- Shipping temperature:4 °C
- Antigen name:Ubiquitin D
- Cat. No.:10749-230
- Reactivity:Human
- Gene ID:O15205
- Antigen synonyms:UBD
- Immunogen:UBD antibody was raised against a 16 amino acid peptide near the center of human UBD. The immunogen is located within amino acids 80 - 130 of UBD.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:UBD
- Storage buffer:UBD antibody is supplied in PBS containing 0.02% sodium azide.
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



