To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:chromosome 12 open reading frame 75
- Antigen symbol:C12orf75
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:0.02 mg
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:Q8TAD7
- Antigen synonyms:C12orf75
- Storage buffer:OCC-1 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:OCC-1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:OCC-1 antibody was raised against a 7 amino acid synthetic peptide near the carboxy terminus of human OCC-1. The immunogen is located within the last 50 amino acids of OCC-1.
- Purification:OCC-1 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10748-782
- Supplier No.:4987_S
Specifications
About this item
OCC-1 Antibody: OCC-1 (Overexpressed in colon carcinoma-1) was initially identified as a novel human cDNA overexpressed in a colon carcinoma cell line. Elevated levels of OCC-1 mRNA were observed in three of eight colon carcinomas, suggesting that OCC-1 may be a hallmark of only a subset of colon carcinomas. It is highly expressed in skeletal muscle, kidney, placenta, and pancreas tissues, with low to no expression in heart, brain, lung and liver tissues. At least two different OCC-1 mRNA species are known to exist.
Type: Primary
Antigen: C12orf75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human