Order Entry
Canada
ContactUsLinkComponent
 
PrimaCell™, Mouse Glomerular Endothelial Cell Culture Kit, CHI Scientific
Catalog # 10783-636
Supplier:  CHI Scientific
CAS Number:  
undefined
PrimaCell™, Mouse Glomerular Endothelial Cell Culture Kit, CHI Scientific
Catalog # 10783-636
Supplier:  CHI Scientific
Supplier Number:  2-82056
CAS Number:  

Specifications

  • Description:
    Mouse Kidney PrimaCell™ 1: Normal Glomerular Endothelial Cells
  • Size:
    Kit
  • Cat. no.:
    10783-636
  • Supplier No.:
    2-82056

Specifications

About this item

The PrimaCell™ system has been developed for the acquisition and growth of primary cells from a variety of different tissue types. Each PrimaCell™ kit has been optimized for each cell type to produce 4-7 times the number of primary cells obtained from published literature protocols. Each kit comes with 100 ml of tissue washing medium, our optimized tissue dissociation system OptiTDS™, 500 ml of growth medium, and enough growth supplements and serum to add to the supplied medium. Kits that require a Fibroblast control system also come with our FibrOut™ kit (added to the growth medium) which reduces or eliminates Fibroblast growth after 2-3 cell growth cycles.

The PrimaCell™ system has been developed for the acquisition and growth of primary cells from a variety of different tissue types

Stability/Storage: The reagents contained in this system are packed in dry ice or gel ice packs for shipping but the kit should be used immediately or the package should be stored at -20°C upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20°C.

This kit includes (and is composed of) all of the items listed under the Accessories Section. Individual accessory items may be purchased instead of the full kit or in addition to the full kit as desired.

Each PrimaCell™ kit has been optimized for each cell type to produce 4-7 times the number of primary cells obtained from published literature protocols. Each kit comes with 100 ml of tissue washing medium, our optimized tissue dissociation system OptiTDS™, 500 ml of growth medium, and enough growth supplements and serum to add to the supplied medium. Kits that require a Fibroblast control system also come with our FibrOut™ kit (added to the growth medium) which reduces or eliminates Fibroblast growth after 2-3 cell growth cycles.