Orders LinkContactUsLinkComponent
ClipperMop™ Cleanroom Mops
ClipperMop™ Cleanroom Mops
Supplier:   Texwipe
ClipperMop™ Cleanroom Mops
Supplier:  Texwipe
Certificates
ClipperMop™ Cleanroom Mops
ClipperMop™ Cleanroom Mops
2 Options Available Below
Product Details & Documents
The mop system may be used with wet, dry or sterile wipers, and is compatible with protocols for ISO Class 3 to 8 cleanrooms, sterile and aseptic areas
  • Cleanroom compatible mopping system
  • Patent-pending thermoplastic mop with clips
  • Quick and easy wiper change
  • Lightweight, easy to handle
  • Less expensive than competitors cleanroom mops
The small size conveniently cleans hard-to-reach areas, walls, ceilings, floors and equipment exteriors. The mop system may be sterilized by gamma irradiation, chemicals, vaporized hydrogen peroxide (VHP) or ethylene oxide. Widely used in pharmaceutical, biomedical, aseptic pharmaceutical compounding and medical device industries.
The patent-pending mophead utilizes non-abrasive polyethylene clips to securely fasten one or multiple wipers to the head. The ClipperMop wiper attachment method does not release particles, fibers and dirt.
Ordering information: The ClipperMop™ cleanroom mop includes the Celcon® thermoplastic mop head with clips, a flexible polyester pad for better wiping efficiency and an adjustable handle.
Documents
Product Family Options
Product Information
  • Dimensions
    Availability
    Price
  • TWTX7102
    ClipperMop™ Cleanroom mop with head assembly and telescoping handle
    ClipperMop™ Cleanroom mop with head assembly and telescoping handle
    18 x 10 cm (7 x 4")
    Log in for availability
    Log in for availability
    Each
    Each - $521.90
    $521.90
    New Customer Promotional Price
    Each
       $521.90
    Supplier Number: TX7102
  • TWTX7103
    ClipperMop™ Cleanroom mop with head assembly and telescoping handle
    ClipperMop™ Cleanroom mop with head assembly and telescoping handle
    28 x 10 cm (11 x 4")
    Log in for availability
    Log in for availability
    Each
    Each - $543.63
    $543.63
    New Customer Promotional Price
    Each
       $543.63
    Supplier Number: TX7103
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • ClipperMop™ tether clip replacement kit, 4 polyethylene tether clips and 2 nylon screws per bag
    ClipperMop™ tether clip replacement kit, 4 polyethylene tether clips and 2 nylon screws per bag
    TWTX7102C
    Log in for availability
    Case of 20 - $322.04
Product Information
  • Catalog #
    Availability
    Price
  • ClipperMop™ replacement pads, foam pads (101,6×177,8) and white fasteners for use with TX7103
    ClipperMop™ replacement pads, foam pads (101,6×177,8) and white fasteners for use with TX7103
    TWTX7111B
    Log in for availability
    Bag of 10 - $13.58