OmniPur® Calbiochem® DNA Extraction Buffer, MilliporeSigma
CA-EM3700
:
- Concentration:225 - 260 mM
- Size:20 L
- Storage temperature:+15°C to +30°C
- Cat. no.:CA-EM3700
- pH:6.9 - 7.1 (25°C)
- Supplier No.:3700
- Conductivity at 25 °C: 35 - 55 mS
- DNase: None detected