Order Entry
Canada
ContactUsLinkComponent
Human Recombinant CD74 (from HEK293 Cells)
Human Recombinant CD74 (from HEK293 Cells)
Catalog # 10797-164
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant CD74 (from HEK293 Cells)
Catalog # 10797-164
Supplier:  Prosci
Supplier Number:  96-110
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.1 mg
  • Tag sequence:
    His Tag
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilized human CD74 at 5μg/ml ( 100μl/well ) can bind biotinylated human CTSL1 with a linear range of 3. 2 - 500 ng/mL.
  • Protein synonyms:
    HLADG|II|INVG34|CLIP|p33|CD74|DHLAG|Ia-GAMMA
  • Protein/peptide name:
    CD74
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    19.3 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot
  • Cat. no.:
    10797-164
  • Supplier No.:
    96-110

Specifications

About this item

Cluster of Differentiation 74 (CD74) is also known as HLA class II histocompatibility antigen gamma chain (HLADG), DHLAG, Ia-GAMMA, invariant chain (Abbreviated Ii). CD74 / HLADG is a polypeptide involved in the formation and transport of MHC class II protein. The cell surface form of the invariant chain is known as CD74. CD74 is upregulated in several cancers and is also expressed by non-immune cells during inflammation. CD74 plays a critical role in MHC class II antigen processing by stabilizing peptide-free class II alpha / beta heterodimers in a complex soon after their synthesis and directing transport of the complex from the endoplasmic reticulum to the endosomal / lysosomal system where the antigen processing and binding of antigenic peptides to MHC class II takes place. CD74 / DHLAG also serves as cell surface receptor for the cytokine MIF.