Human Recombinant BMPR1A (from HEK293 cells)
Catalog # 10796-980
Supplier: Prosci
CAS Number:
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.2 mg
- Tag sequence:His Tag
- Protein synonyms:BMPR-1A|SKR5|ALK-3|10q23del|CD292|ACVRLK3
- Protein/peptide name:BMPR1A
- Purity:>97% as determined by SDS-PAGE.
- Molecular weight:15.7 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot
- Cat. no.:10796-980
- Supplier No.:96-023
Specifications
About this item
The bone morphogenetic protein receptor, type IA is also known as BMPR1A or ALK3 is a protein which in humans is encoded by the BMPR1A gene. BMPR1A has also been designated as CD292 (cluster of differentiation 290). The bone morphogenetic protein (BMP) receptors are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A (this protein) and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. Alk3-mediated BMP signaling in AV endocardial/mesenchymal cells plays a central role during cushion morphogenesis.