dNTP Mix, 10mM, MilliporeSigma
Catalog # CA80500-356
Supplier: MilliporeSigma
CAS Number:
Specifications
- Description:dNTP Mix
- Size:0.2 mL
- Storage conditions:Store at -20° C
- Concentration:10 mM
- Cat. no.:CA80500-356
- Supplier No.:71004-3
Specifications
About this item
The dNTP Mix contains ultrapure dATP, dCTP, dGTP, and dTTP (monosodium salts) at a concentration of 10 mM each in sterile deionized water at pH 7.0.
- Ready to use
- RNase and DNase free
Ideal for any application that requires pure deoxynucleotides, such as PCR, cDNA synthesis, and fill-in reactions.