Anti-DRAK2 (serine/threonine kinase 17b) Goat Polyclonal Antibody
89296-346
: Genetex
- Antibody type:Primary
- Antigen name:Serine/Threonine Kinase 17B
- Antigen symbol:DRAK2 (serine/threonine kinase 17b)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Dog
- Western blot:Yes
- Size:100 μg
- Gene ID:9262
- Antigen synonyms:DRAK2
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-HSVRSSEDKTSK
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89296-346
- Supplier No.:GTX88405
Goat polyclonal antibody to DRAK2
Approx 45kDa band observed in lysates of cell line MOLT4 (calculated MW of 42.3kDa according to NP_004217.1). Recommended concentration: 1-3µg/ml. This antiboyd may cross-react with rat STK17B (GeneID: 170904).
Type: Primary
Antigen: DRAK2 (serine/threonine kinase 17b)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Dog