Anti-STK39 Rabbit Polyclonal Antibody
Catalog # 89328-834
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Serine/Threonine Kinase 39
- Antigen symbol:STK39
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 μg
- Gene ID:27347
- Antigen synonyms:PASK|SPAK|DCHT|Ste-20-related kinase
- Storage buffer:Antibody is supplied in PBS containing 0.02% sodium azide
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Purification:Affinity chromatography purified via peptide column
- Cat. no.:89328-834
- Supplier No.:GTX85023
Specifications
About this item
Rabbit polyclonal antibody to Stk39
Type: Primary
Antigen: STK39
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat