Anti-CD36 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TR9]
89298-734
: Genetex
- Antibody type:Primary
- Antigen name:CD36 molecule
- Antigen symbol:CD36
- Clonality:Monoclonal
- Clone:TR9
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 test
- Gene ID:948
- Antigen synonyms:GP4|GP3B|FAT|GPIV|SCARB3|CHDS7|BDPLT10|PASIV
- Storage buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage temperature:Store in the dark at 2-8°C. Do not freeze. Do not use after expiration date stamped on vial label. Short-term exposure to room temperature should not affect the quality of the reagent.
- Immunogen:Platelets
- Cat. no.:89298-734
- Supplier No.:GTX79880
FITC-conjugated Mouse Monoclonal antibody [TR9] to CD36
Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: TR9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: